Human recombinant Lecithin-cholesterol acyltransferase (from cells)
Fournisseur: ProSci Inc.
91-786
PRSI91-786EA 678 EUR InStock
PRSI91-786
Human recombinant Lecithin-cholesterol acyltransferase (from cells)
Enzymes
Lipase family. The gene encoding this protein is expressed mainly in brain, liver and testes,followed by secreting into plasma and cerebral spinal fluid. The esterification of cholesterol is required for cholesterol transport. LCAT is a central enzyme in the extracellular metabolism of plasma lipoproteins. Defects in LCAT are the cause of lecithin-cholesterol acyltransferase deficiency (LCATD) and a cause of fish-eye disease (FED).
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité