Human recombinant IL-1RAcP (from cells)
Fournisseur: ProSci Inc.
91-787
PRSI91-787EA 306 EUR InStock
PRSI91-787
Human recombinant IL-1RAcP (from cells)
Protéines et Peptides
Interleukin-1 Receptor Accessory Protein (IL-1RAcP) is a member of the interleukin-1 receptor family. It contains three Ig-like C2-type domains in the extracellular region and a long cytoplasmic domain implicated in signal transduction. IL-1RAcP acts as a non-ligand binding accessory component of the receptors for IL1 alpha, IL1 beta and IL33. IL-1RAcP mediates interleukin-1-dependent activation of NF-kappa-B. It is part of the membrane-bound form of the IL-1 receptor. IL-1 RAcP takes part in the Signaling ways by the formation of a ternary complex containing IL1R1, TOLLIP, MYD88, and IRAK1 or IRAK2. In addition, IL-1RAcP modulates the response to interleukins by associating with soluble IL1R1 and enhancing interleukin-binding to the decoy receptor.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité