Human Recombinant SOD2 (from Cells)
Fournisseur: ProSci Inc.
91-789
PRSI91-789EA 588 EUR InStock
PRSI91-789
Human Recombinant SOD2 (from Cells)
Protéines et Peptides
Superoxide Dismutase (SOD2) belongs to the iron/manganese superoxide dismutase family. SOD2 is a mitochondrial matrix protein that forms a homotetramer and binds one manganese ion per subunit. SOD2 transforms toxic superoxide, a byproduct of the mitochondrial electron transport chain into hydrogen peroxide and diatomic oxygen. It is reported that oxidative stress plays an essential role in the development of breast cancer, while SOD2 is one of the primary enzymes that directly convert potential harmful oxidizing species to harmless metabolites.
  • Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays.
Attention: For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité