Human recombinant Alkaline Phosphatase (from cells)
Fournisseur: ProSci Inc.
91-795
PRSI91-795EA 789 EUR InStock
PRSI91-795
Human recombinant Alkaline Phosphatase (from cells)
Enzymes
ALPP is a membrane protein and exits as a homodimer. ALPP is expressed only in normal term placenta, endocervix and fallopian tube and also in ovarian and proximal gastrointestinal tumors. It has been shown to play a role in a number of processes including cell signaling, long-term potentiation, and cell adhesion, however, the best known and most commonly studied role is implicated in Alzheimer's research.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité