Human recombinant IL-2 R gamma (from cells)
Fournisseur: ProSci Inc.
91-799
PRSI91-799EA 588 EUR InStock
PRSI91-799
Human recombinant IL-2 R gamma (from cells)
Protéines et Peptides
IL2RG contains one fibronectin type-III domain. IL2RG is an important signaling component of many interleukin receptors, including those of interleukin -2, -4, -7 and -21, and is thus referred to as the common gamma chain. IL2RG interacts with SHB upon interleukin stimulation and HTLV-1 accessory protein p12I. Defects in IL2RG are the cause of X-linked combined immunodeficiency (XCID) and severe combined immunodeficiency X-linked T-cell-negative /B-cell-positive / NK-cell-negative (XSCID).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité