Human recombinant Apolipoprotein C-II (from E. coli)
Fournisseur: ProSci Inc.
91-800
PRSI91-800EA 884 EUR InStock
PRSI91-800
Human recombinant Apolipoprotein C-II (from E. coli)
Protéines et Peptides
APOC2 activates the lipoprotein lipase in capillaries, which hydrolyzes triglycerides and thus provides free fatty acids for cells. APOC2 is component of the very low density lipoprotein (VLDL) fraction in plasma. It is also an activator of several triacylglycerol lipases. The association of APOC2 with plasma chylomicrons, VLDL, and HDL is reversible, a function of the secretion and catabolism of triglyceride-rich lipoproteins, and changes rapidly. Defects in APOC2 are the cause of hyperlipoproteinemia type 1B (HLPP1B) which characterized by hypertriglyceridemia, xanthomas, and increased risk of pancreatitis and early atherosclerosis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité