Human recombinant LRG1 (from cells)
Fournisseur: ProSci Inc.
91-805
PRSI91-805EA 654 EUR InStock
PRSI91-805
Human recombinant LRG1 (from cells)
Protéines et Peptides
LRG1 is a secreted protein and contains eight LRR repeats and one LRRCT domain, It can be expressed during granulocyte differentiation. LRG1 has been shown to be involved in protein-protein interaction, signal transduction, cell adhesion and development. Levels of the LRG protein are markedly elevated in acute appendicitis and therefore could be used as a diagnostic aid.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité