Human recombinant R-spondin 3 (from cells)
Fournisseur: ProSci Inc.
91-806
PRSI91-806EA 727 EUR InStock
PRSI91-806
Human recombinant R-spondin 3 (from cells)
Protéines et Peptides
R-spondin-3 (RSPO3), also known as Protein with TSP type-1 repeat, Roof plate-specific spondin-3, Thrombospondin type-1 domain-containing protein 2, PWTSR, THSD2 and CRISTIN1, is a member of the thrombospondin type 1 repeat supergene family. RSPO3 is a secreted protein and widely expressed in many tissues. RSPO3 contains two Furin-like repeats which have been found in a variety of eukaryotic proteins involved in the mechanism of signal transduction by receptor tyrosine kinases, and one TSP type-1 domain, RSPO3 founctions as a activator of the beta-catenin signaling cascade, leading to TCF-dependent gene activation. Otherwise, RSPO3 may negatively regulate the TGF-beta pathway.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité