Human recombinant CYR61 (from cells)
Fournisseur: ProSci Inc.
91-813
PRSI91-813EA 588 EUR InStock
PRSI91-813
Human recombinant CYR61 (from cells)
Protéines et Peptides
Protein CYR61, also known as CCN family member 1, Cysteine-rich angiogenic inducer 61,Insulin-like growth factor-binding protein 10 , GIG1, CYR61, CCN1 and IGFBP10, belongs to the CCN family, CYR61 is a secreted protein and contains one CTCK (C-terminal cystine knot-like) domain,one IGFBP N-terminal domain,one TSP type-1 domain and one VWFC domain. CYR61 promotes cell proliferation, chemotaxis, angiogenesis and cell adhesion. CYR61 plays important roles in inflammation and tissue repair. CYR61 is associated with diseases related to chronic inflammation, including rheumatoid arthritis, atherosclerosis, diabetes-related nephropathy and retinopathy, and many different forms of cancers.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité