Human recombinant IL-18 binding (from cells)
Fournisseur: ProSci Inc.
91-814
PRSI91-814EA 343 EUR InStock
PRSI91-814
Human recombinant IL-18 binding (from cells)
Protéines et Peptides
Interleukin-18-binding protein, also known as Tadekinig-alfa and IL18BP, contains 1 Ig-like C2-type domain. IL18BP is constitutively expressed and secreted in mononuclear cells. IL18BP functions as an IL18 inhibitor. IL18BP binds to IL18, prevents the binding of IL18 to its receptor, and thus inhibits IL18-induced IFN-gamma production. It has been shown that IL18BP may be a promising molecular approach to inhibit neointimal hyperplasia and arteriosclerosis progression following coronary and peripheral angioplasty.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité