Human recombinant neuronal cell adhesion molecule (from cells)
Fournisseur: ProSci Inc.
91-816
PRSI91-816EA 588 EUR InStock
PRSI91-816
Human recombinant neuronal cell adhesion molecule (from cells)
Protéines et Peptides
Neuronal cell adhesion molecule(NRCAM) is a single-pass type I membrane protein ,containing 5 fibronectin type-III domains and 6 Ig-like C2-type (immunoglobulin-like) domains.It belongs to the immunoglobulin superfamily. NrCAM is engaged in such biological processes as axonal fasciculation, cell-cell adhesion, central nervous system development, clustering of voltage-gated sodium channels, neuron migration, positive regulation of neuron differentiation, regulation of axon extension, and synaptogenesis. It also may play a role in the molecular assembly of the nodes of Ranvier. NrCAM effects are also linked with different recognition processes and signal transduction pathways regulating cell differentiation, proliferation, or migration
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité