Mouse recombinant FLT3 ligand
Fournisseur: ProSci Inc.
91-820
PRSI91-820EA 654 EUR InStock
PRSI91-820
Mouse recombinant FLT3 ligand
Protéines et Peptides
Fms-related tyrosine kinase 3 ligand(Flt3L) is a single-pass type I membrane protein and consists of 232 amino acids. Flt3L is a hematopoietic four helical bundle cytokine, structurally homologous to stem cell factor and colony stimulating factor. Flt3L synergizes well with a number of other colony stimulating factors and interleukins. Flt3L stimulates the proliferation and differentiation of various blood cell progentiors by activating FLT3.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité