Mouse recombinant cathepsin D
Fournisseur: ProSci Inc.
91-821
PRSI91-821EA 761 EUR InStock
PRSI91-821
Mouse recombinant cathepsin D
Protéines et Peptides
CTSD localizes to the lysosome and consists of a light chain and a heavy chain. CTSD is expressed in epithelial cells as well as in macrophages.CTSD is a lysosomal aspartyl protease that depends critically on protonation of its active site Asp residue and gets activated at pH 5 in endosome of hepatocytes. It has been suggested to facilitate cancer cell migration and invasion by digesting the basement membrane, extracellular matrix and xonnective tissue. In addition, CTSD has been used as a breast cancer tumor marker.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité