Human recombinant IFN alpha 4 (from cells)
Fournisseur: ProSci Inc.
91-822
PRSI91-822EA 588 EUR InStock
PRSI91-822
Human recombinant IFN alpha 4 (from cells)
Protéines et Peptides
Interferon stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase. They allow for communication between cells to trigger the protective defenses of the immune system that eradicate pathogens or tumors. They are typically divided among three IFN classes: Type I, Type II and Type III. IFNA4 is a secreted protein and produced by macrophages. Two variants of IFNA4 (IFNA4a and IFNA4b) are known, which differ from each other by changes in their coding regions at nucleotide positions 220 and 410 .
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité