Human recombinant lymphotactin (from cells)
Fournisseur: ProSci Inc.
91-823
PRSI91-823EA 654 EUR InStock
PRSI91-823
Human recombinant lymphotactin (from cells)
Protéines et Peptides
Lymphotactin is a secreted protein which belongs to the intercrine gamma family. It is also a member of the XC chemokine family. XCL1 is found in spleen with highest level, lower in peripheral leukocytes and very low levels in lung, colon and small intestine. XCL1 plays a role in inflammatory and immunological responses, inducing leukocyte migration and activation. XCL1 induces chemotactic function by binding to a chemokine receptor called XCR1. XCL1 is closely related to another chemokine called XCL2.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité