Human recombinant SEMA4B (from cells)
Fournisseur: ProSci Inc.
91-824
PRSI91-824EA 852 EUR InStock
PRSI91-824
Human recombinant SEMA4B (from cells)
Protéines et Peptides
Semaphorin-4B is a single-pass type I membrane protein. SEMA4B is a member of the semaphorin family. The class 4 semaphorins are integral membrane proteins that are widely expressed throughout the nervous system. SEMA4B is a single-pass type I membrane protein and contains one Ig-like C2-type (immunoglobulin-like) domain, one PSI domain and one Sema domain. Human SEMA4B is expressed in neurons. SEMA4B inhibits axonal extension by providing local signals to specify territories inaccessible for growing axons. SEMA4B negatively regulates basophil-mediated Th2 and humoral memory responses.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité