Human Recombinant CCL24 (from Cells)
Fournisseur: ProSci Inc.
91-830
PRSI91-830EA 620 EUR InStock
PRSI91-830
Human Recombinant CCL24 (from Cells)
Protéines et Peptides
Cytokine are a family of secreted proteins involved in immunoregulatory and inflammatory processes. The CC cytokines are proteins characterized by two adjacent cysteine. CCL24 diaplays the chemotactic for resting T-lymphocytes and eosinophils. CCL24 has lower chemotatic activity for neutrophils, none for monocytes and activated lymphocytes. CCL24 is a strong suppressor of colony formation by a multipotential hematopoietic progenitor cell lines.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité