Mouse recombinant CXCL16
Fournisseur: ProSci Inc.
91-831
PRSI91-831EA 654 EUR InStock
PRSI91-831
Mouse recombinant CXCL16
Protéines et Peptides
CXCL16 is a single-pass type I membrane protein, which consists of 246 amino acids, CXCL16 induces a strong chemotatic response and calcium mobilization. CXCL16 acts as a scavenger receptor on macrophages, which specially binds to oxidized low density lipoprotein. CXCL16 may involves in pathophysiology such as atherogenesis. Soluble CXCL16 may play an important role in liver metastases through the induction of epithelial-mesenchymal transition.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité