Human recombinant IL21 (fromE. coli)
Fournisseur: ProSci Inc.
91-832
PRSI91-832EA 852 EUR InStock
PRSI91-832
Human recombinant IL21 (fromE. coli)
Protéines et Peptides
IL-21 induces the production of IgG1 and IgG3 in B-cells. IL-21 may promote the transition between innate and adaptive immunity. IL-21 may have a role in proliferation and maturation of natural killer cells in synergy with IL15. IL-21 stimulates interferon gamma production in T-cells and natural killer cells by synergy with IL15 and IL18. Dysregulation of IL-21 has a role in multiple immune-mediated diseases including lupus, psoriasis and chronic inflammatory diseases.
  • This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité