Human recombinant Dickkopf-related 2 (from cells)
Fournisseur: ProSci Inc.
91-834
PRSI91-834EA 654 EUR InStock
PRSI91-834
Human recombinant Dickkopf-related 2 (from cells)
Protéines et Peptides
Dickkopf-related protein 2 (DKK2), is a member of the dickkopf family. DKK2 is secreted protein which contains two cysteine rich regions and is involved in embryonic development through its interactions with the Wnt signaling pathway. It can act as either an agonist or antagonist of Wnt/beta-catenin signaling, depending on the cellular context and the presence of the co-factor kremen 2. It antagonizes canonical Wnt signaling by inhibiting LRP5/6 interaction with Wnt and by forming a ternary complex with the transmembrane protein KREMEN that promotes internalization of LRP5/6. DKKs play an important role in vertebrate development, where they locally inhibit Wnt regulated processes such as antero-posterior axial patterning, limb development, somitogenesis and eye formation. In the adult, Dkks are implicated in bone formation and bone disease, cancer and Alzheimer disease.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: N-Fc, C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité