Mouse recombinant Ephrin A4
Fournisseur: ProSci Inc.
91-835
PRSI91-835EA 343 EUR InStock
PRSI91-835
Mouse recombinant Ephrin A4
Protéines et Peptides
Ephrin-A4 belongs to the ephrin family and Contains 1 ephrin RBD (ephrin receptor-binding) domain. The protein is cell surface GPI-bound ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. Ephrin-A4 binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. It may play a role in the interaction between activated B-lymphocytes and dendritic cells in tonsils.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité