Human recombinant Ephrin B1 (from cells)
Fournisseur: ProSci Inc.
91-837
PRSI91-837EA 620 EUR InStock
PRSI91-837
Human recombinant Ephrin B1 (from cells)
Protéines et Peptides
Ephrin-B1, also named EFL-3, ELK ligand, EPH-related receptor tyrosine kinase ligand 2, is a single-pass type I membrane protein. It contains 1 ephrin RBD (ephrin receptor-binding) domain and belongs to the ephrin family. Ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. All ephrins share a conserved extracellular sequence, which most likely corresponds to the receptor-binding domain. Ephrin-B1 has been shown to bind EphA3, EphB1, EphB2, EphB3, and EphB4. The extracellular domains of human and mouse ephrin-B1 share 94% amino acid identity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité