Human recombinant Carboxypeptidase M (from Cells)
Fournisseur: ProSci Inc.
91-841
PRSI91-841EA 884 EUR InStock
PRSI91-841
Human recombinant Carboxypeptidase M (from Cells)
Enzymes
These enzymes remove C-terminal amino acids from peptides and proteins and exert roles in the physiological processes of blood coagulation/fibrinolysis, inflammation, food digestion and pro-hormone and neuropeptide processing. CPM is widely distributed in a variety of tissues and cells. CPM is involved in peptide metabolism on both the cell surface and in extracellular fluids. CPM functions not only as a protease but also as a binding partner in cell-surface protein-protein interactions.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité