Human recombinant BPI (from Cells)
Fournisseur: ProSci Inc.
91-842
PRSI91-842EA 525 EUR InStock
PRSI91-842
Human recombinant BPI (from Cells)
Protéines et Peptides
Bactericidal permeability-increasing protein(BPI for short), is a secreted protein which belongs to the BPI/LBP/Plunc superfamily, BPI/LBP family. It exists as a monomer or a disulfide-linked homodimer. The cytotoxic action of BPI is limited to many species of Gram-negative bacteria. This specificity may be explained by a strong affinity of the very basic N-terminal half for the negatively charged lipopolysaccharides that are unique to the Gram-negative bacterial outer envelope. BPI has antibacterial activity against the Gram-nagative bacterium P.aeruginosa, and this activity is inhibited by LPS from P.aeruginosa.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité