Human recombinant VCP-like (from cells)
Fournisseur: ProSci Inc.
91-847
PRSI91-847EA 884 EUR InStock
PRSI91-847
Human recombinant VCP-like (from cells)
Protéines et Peptides
Probable serine carboxypeptidase CPVL, also known as Carboxypeptidase, vitellogenic-like, Vitellogenic carboxypeptidase-like protein, is a member of the peptidase S10 family. It is expressed in macrophages but not in other leukocytes. And specifically, it is abundantly expressed in heart and kidney, also expressed in spleen, leukocytes, and placenta. This enzyme may be involved in the digestion of phagocytosed particles in the lysosome, and also participation in an inflammatory protease cascade, and trimming of peptides for antigen presentation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité