Human recombinant VIP peptide (from cells)
Fournisseur: ProSci Inc.
91-856
PRSI91-856EA 654 EUR InStock
PRSI91-856
Human recombinant VIP peptide (from cells)
Protéines et Peptides
Vasoactive intestinal peptide is also known as the vasoactive intestinal polypeptide or VIP. In humans, it is encoded by the VIP gene. VIP is neuropeptide which belongs to a glucagon/secretin superfamily, the ligand of class II G protein-coupled receptors. VIP is produced in many tissues of vertebrates including the gut, pancreas and suprachiasmatic nuclei of the hypothalamus in the brain. VIP stimulates contractility in the heart, causes vasodilation, lowers arterial blood pressure and relaxes the smooth muscle of trachea, stomach and gall bladder. VIP has a half-life (t½) in the blood of about two minutes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité