Human recombinant SEMA4D (from cells)
Fournisseur: ProSci Inc.
91-858
PRSI91-858EA 553 EUR InStock
PRSI91-858
Human recombinant SEMA4D (from cells)
Protéines et Peptides
Semaphorin-4D is also known as A8,BB18, GR3, CD100. Semaphorin-4D belongs to the semaphorin family containing 1 Ig-like C2-type domain, 1 PSI domain and 1 Sema domain. It is the cell surface receptor for PLXN1B and PLXNB2 that plays an important role in cell-cell signaling. It promotes the migration of cerebellar granule cells and of endothelial cells, regulates dendrite and axon branching and morphogenesis. Semaphorin-4D Plays a role in the immune system; Promotes signaling via SRC and PTK2B/PYK2, which then mediates activation of phosphatidylinositol 3-kinase and of the AKT1 signaling cascade.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité