Human recombinant LAMP (from cells)
Fournisseur: ProSci Inc.
91-862
PRSI91-862EA 588 EUR InStock
PRSI91-862
Human recombinant LAMP (from cells)
Protéines et Peptides
Limbic system-associated membrane protein is also known as LSAMP, IgLON family member 3. In humans, it is encoded by the LSAMP gene. It belongs to the immunoglobulin superfamily and contains 3 Ig-like C2-type domains. Limbic system-associated membrane protein mediates selective neuronal growth and axon targeting. It contributes to the guidance of developing axons and remodeling of mature circuits in the limbic system. It is also essential for normal growth of the hyppocampal mossy fiber projection
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité