Human recombinant beta-1,4-Galactosyltransferase 3 (from Cells)
Fournisseur: ProSci Inc.
91-863
PRSI91-863EA 789 EUR InStock
PRSI91-863
Human recombinant beta-1,4-Galactosyltransferase 3 (from Cells)
Enzymes
Beta-1,4-galactosyltransferase 3 (B4GALT3) belongs to the glycosyltransferase 7 family. It is responsible for the synthesis of complex-type N-linked oligosaccharides in many glycoproteins as well as the carbohydrate moieties of glycolipids. It is highest expression in placenta, prostate, testis, ovary, intestine and muscle, and in fetal brain.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité