Human recombinant IL13 (from cells)
Fournisseur: ProSci Inc.
91-866
PRSI91-866EA 852 EUR InStock
PRSI91-866
Human recombinant IL13 (from cells)
Protéines et Peptides
Interleukin-13 is also known as IL-13. It is a protein that in humans is encoded by the IL13 gene. Interleukin-13 is an immunoregulatory cytokine produced primarily by activated Th2 cells.It is involved in several stages of B-cell maturation and differentiation. It up-regulates CD23 and MHC class II expression, and promotes IgE isotype switching of B cells. This cytokine down-regulates macrophage activity, thereby inhibits the production of pro-inflammatory cytokines and chemokines. This cytokine is found to be critical to the pathogenesis of allergen-induced asthma but operates through mechanisms independent of IgE and eosinophils.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité