Human recombinant PS-Beta-G-3 (from cells)
Fournisseur: ProSci Inc.
91-867
PRSI91-867EA 761 EUR InStock
PRSI91-867
Human recombinant PS-Beta-G-3 (from cells)
Protéines et Peptides
Pregnancy-specific beta-1-glycoprotein 3 is also known as Carcinoembryonic Antigen SG5,Pregnancy-Specific Glycoprotein 3 ,PS-Beta-G-3, PSBG-3.It belongs to the immunoglobulin superfamily. CEA family.It synthesized in large amounts by placental trophoblasts and released into the maternal circulation during pregnancy. Molecular cloning and analysis of several PSG genes has indicated that the PSGs form a subgroup of the carcinoembryonic antigen (CEA) gene family, Members of the CEA family consist of a single N domain, with structural similarity to the immunoglobulin variable domains, followed by a variable number of immunoglobulin constant-like A and/or B domains.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité