Human recombinant Butyrophilin 3A2 (from Cells)
Fournisseur: ProSci Inc.
91-868
PRSI91-868EA 761 EUR InStock
PRSI91-868
Human recombinant Butyrophilin 3A2 (from Cells)
Protéines et Peptides
Butyrophilin subfamily 3 member A2, also known as BT3.2, BTF3, BTF4 and BTN3A2, is a single-pass type I membrane protein. It is a member of the butyrophilin (BTN) family and the immunoglobulin (IG) superfamily. BTN3A2 plays a role in T-cell responses in the adaptive immune response and inhibits the release of IFNG from activated T-cells.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité