Human recombinant Follistatin-like 1 (from cells)
Fournisseur: ProSci Inc.
91-871
PRSI91-871EA 525 EUR InStock
PRSI91-871
Human recombinant Follistatin-like 1 (from cells)
Protéines et Peptides
Follistatin-Related Protein 1 (FSTL1) is a secreted protein that contains two EF-hand domains, one follistatin-like domain, one Kazal-like domain, and one VWFC domain. Its functional significance in physiological and pathological processes is not completely understood. However, FSTL1 is thought to modulate the action of some growth factors on cell proliferation and differentiation. FSTL1 maybe an autoantigen associated with rheumatoid arthritis.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité