Human recombinant Creatine Kinase (from cells)
Fournisseur: ProSci Inc.
91-873
PRSI91-873EA 419 EUR InStock
PRSI91-873
Human recombinant Creatine Kinase (from cells)
Enzymes
Creatine kinase M-type is also known as Creatine kinase M chain,M-CK. It is a protein that in humans is encoded by the CKM gene. It belongs to the ATP:guanido phosphotransferase family,containing 1 phosphagen kinase C-terminal domain and 1 phosphagen kinase N-terminal domain. Creatine kinase M-type can reversibly catalyses the transfer of phosphate between ATP and various phosphogens. It plays a central role in energy transduction in tissues with large, fluctuating energy demands, such as skeletal muscle, heart, brain and spermatozoa.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité