Human recombinant CTHRC1 (from cells)
Fournisseur: ProSci Inc.
91-875
PRSI91-875EA 852 EUR InStock
PRSI91-875
Human recombinant CTHRC1 (from cells)
Protéines et Peptides
Collagen triple helix repeat-containing protein 1 is a protein that in humans is encoded by the CTHRC1 gene. It acts as a negative regulator of collagen matrix deposition. It may cause the disease of Barrett esophagus . Patients with Barrett esophagus have an increased risk of esophageal adenocarcinoma. The main cause of Barrett esophagus is gastroesophageal reflux. The retrograde movement of acid and bile salts from the stomach into the esophagus causes prolonged injury to the esophageal epithelium and induces chronic esophagitis, which in turn is believed to trigger the pathologic changes.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité