Human Recombinant CD2 (from Cells)
Fournisseur: ProSci Inc.
91-876
PRSI91-876EA 852 EUR InStock
PRSI91-876
Human Recombinant CD2 (from Cells)
Protéines et Peptides
T-cell surface antigen CD2 is also known as Erythrocyte receptor, LFA-2, LFA-3 receptor, Rosette receptor, T-cell surface antigen T11/Leu-5. It is a protein that in humans is encoded by the CD2 gene. It is a single-pass type I membrane protein. contains 1 Ig-like C2-type domain and 1 Ig-like V-type domain. T-cell surface antigen CD2 interacts with lymphocyte function-associated antigen (LFA-3) and CD48/BCM1 to mediate adhesion between T-cells and other cell types. It is implicated in the triggering of T-cells, the cytoplasmic domain is implicated in the signaling function.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité