Human recombinant Ephrin Type-B Receptor 1 (from cells)
Fournisseur: ProSci Inc.
91-886
PRSI91-886EA 852 EUR InStock
PRSI91-886
Human recombinant Ephrin Type-B Receptor 1 (from cells)
Protéines et Peptides
Ephrin Type-B Receptor 1 (EPHB1) is a single-pass type I membrane protein that belongs to the Ephrin-B family of receptor tyrosine kinases involved in the development of embryonic nervous and vascular systems. EPHB1 contains two fibronectin type-III domains, one protein kinase domain and one Sterile Alpha Motif (SAM)domain. EPHB1 is able to stimulate fibroblast motility on extracellular matrix in a kinase-dependent manner, which is also correlated with its association with Grb7, an adaptor molecule implicated in the regulation of cell migration. It binds to Ephrin-B1, Ephrin-B2 and Ephrin-B3. EPHB1 plays an important roles in diverse biological processes including nervous system development, angiogenesis, and neural synapsis formation and maturation and may be involved in cell-cell interactions in the nervous system.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité