Mouse recombinant VEGFD
Fournisseur: ProSci Inc.
91-892
PRSI91-892EA 584 EUR InStock
PRSI91-892
Mouse recombinant VEGFD
Protéines et Peptides
Mouse vascular endothelial growth factor D, (VEGFD) is a member of the platelet-derived growth factor/vascular endothelial growth factor (PDGF/VEGF) family. VEGFD is a secreted protein and highly expressed in fetal and adult lung. It undergoes a complex proteolytic maturation, generating multiple processed forms that bind and activate VEGFR-2 and VEGFR-3 receptors. The structure and function of this protein is similar to VEGFC. VEGFD is growth factor which active in angiogenesis, lymphangiogenesis, and endothelial cell growth, stimulating their proliferation and migration and also has effects on the permeability of blood vessels.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité