Human recombinant somatoliberin (from cells)
Fournisseur: ProSci Inc.
91-893
PRSI91-893EA 343 EUR InStock
PRSI91-893
Human recombinant somatoliberin (from cells)
Protéines et Peptides
Growth-hormone-releasing hormone (GHRH) is a releasing hormone for growth hormone. It is a 44-amino acid peptide hormone produced in the arcuate nucleus of the hypothalamus. GHRH first appears in the human hypothalamus between 18 and 29 weeks of gestation, which corresponds to the start of production of growth hormone and other somatotropes in fetuses. GRF is released by the hypothalamus and acts on the adenohypophyse to stimulate the secretion of growth hormone.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité