Mouse recombinant cathepsin L
Fournisseur: ProSci Inc.
91-898
PRSI91-898EA 852 EUR InStock
PRSI91-898
Mouse recombinant cathepsin L
Protéines et Peptides
Mouse Cathepsin L is a lysosomal cysteine proteinase which is a member of the peptidase C1 family. This protein is a dimer composed of disulfide-linked heavy and light chains, both produced from a single protein precursor. Cathepsin L plays a major role in intracellular protein catabolism. Its substrates include collagen and elastin, as well as alpha-1 protease inhibitor, a major controlling element of neutrophil elastase activity. Cathepsin L has been implicated in several pathologic processes, including myofibril necrosis in myopathies and in myocardial ischemia, and in the renal tubular response to proteinuria.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité