Human recombinant REG4 (from cells)
Fournisseur: ProSci Inc.
91-903
PRSI91-903EA 588 EUR InStock
PRSI91-903
Human recombinant REG4 (from cells)
Protéines et Peptides
REG4 is a secreted contains one C-type lectin domain, high expressed in the gastrointestinal tract, including jejunum, ileum, appendix, pancreas and small intestine. REG4 can be up-regulated by mucosal injury from active Crohn’s disease or ulcerative colitis. In the acid environment, REG4 can maintain carbohydrate recognition activity. REG4 may be involved in inflammatory and metaplastic response of the gastrointestinal epithelium.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité