Human recombinant CXCL10 (from cells)
Fournisseur: ProSci Inc.
91-905
PRSI91-905EA 620 EUR InStock
PRSI91-905
Human recombinant CXCL10 (from cells)
Protéines et Peptides
Human C-X-C Motif Chemokine Ligand 10 (CXCL10) is a non-ELR chemokine secreted by various cell types, such as monocytes, endothelial cells and fibroblasts, in response to IFN- gamma . CXCL10 functions via chemokine receptor CXCR3. CXCL10 has been attributed to several roles, such as chemoattraction for activated T-lymphocytes, inhibition of angiogenesis, and antitumor activity.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité