Human recombinant Clusterin (from cells)
Fournisseur: ProSci Inc.
91-906
PRSI91-906EA 525 EUR InStock
PRSI91-906
Human recombinant Clusterin (from cells)
Protéines et Peptides
Clusterin is a secreted protein which belongs to the Clusterin family. Clusterin is expressed in adult testis, heart, ovary, adrenal gland, brain and liver. Clusterin has been suggested to be involved in several basic biological events such as cell death, tumor progression, and neurodegenerative disorders. In addition,Clusterin is up/ down regulated on the mRNA or protein level in many pathological and clinically relevant situations including cancer, organ regeneration, infection, Alzheimer disease, retinitis pigmentosa, myocardial infarction, renal tubular damage, autoimmunity and others.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité