Human recombinant Sclerostin (from cells)
Fournisseur: ProSci Inc.
91-919
PRSI91-919EA 852 EUR InStock
PRSI91-919
Human recombinant Sclerostin (from cells)
Protéines et Peptides
Sclerostin, also known as SOST, is a member of the Cerberus/DAN family of BMP antagonists. SOST is asecreted glycoprotein with a C-terminal cysteine knot-like (CTCK) domain. It shows sequence similarity to the DAN (differential screening-selected gene aberrative in neuroblastoma) family of bone morphogenetic protein (BMP) antagonists. SOST is produced by the osteocyte and has anti-anabolic effects on bone formation. It is a negative regulator of bone growth that acts through inhibition of Wnt signaling and bone formation.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité