Human recombinant PI3K interacting 1 (from cells)
Fournisseur: ProSci Inc.
91-922
PRSI91-922EA 525 EUR InStock
PRSI91-922
Human recombinant PI3K interacting 1 (from cells)
Protéines et Peptides
Phosphoinositide-3-kinase-interacting protein 1(PIK3IP1) is an enzyme that in humans is encoded by the PIK3IP1 gene.It is a negative regulator of phosphatidylinositol-3-kinase (PI3K), suppresses the development of hepatocellular carcinoma. The gene encoding PIK3IP1 maps to human chromosome 22, which houses over 500 genes and is the second smallest human chromosome. Mutations in several of the genes that map to chromosome 22 are involved in the development of Phelan-McDermid syndrome, Neurofibromatosis type 2, autism and schizophrenia.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité