Human recombinant Endoplasmic Reticulum Resident 44 (from cells)
Fournisseur: ProSci Inc.
91-923
PRSI91-923EA 654 EUR InStock
PRSI91-923
Human recombinant Endoplasmic Reticulum Resident 44 (from cells)
Protéines et Peptides
Endoplasmic reticulum resident protein 44 (TXNDC4) is is a 406 amino acid protein that contains one thioredoxin domain. TXNDC4 mediates thiol-dependent retention in the early secretory pathway and forms mixed disulfides with substrate proteins through its conserved CRFS motif.It can inhibit the calcium channel activity of ITPR1.It may have a role in the control of oxidative protein folding in the endoplasmic reticulum.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité