Human recombinant alpha-1-Acid Glycoprotein 2 (from cells)
Fournisseur: ProSci Inc.
91-925
PRSI91-925EA 588 EUR InStock
PRSI91-925
Human recombinant alpha-1-Acid Glycoprotein 2 (from cells)
Protéines et Peptides
Alpha-1-acid glycoprotein 2 is a secreted protein that belongs to the calycin superfamily, lipocalin family.It is expressed by the liver and secreted in plasma. It appears to function in modulating the activity of the immune system during the acute-phase reaction. It functions as transport protein in the blood stream. It binds various hydrophobic ligands in the interior of its beta-barrel domain. It also binds synthetic drugs and influences their distribution and availability.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité