Human recombinant BMP Receptor IA (from Cells)
Fournisseur: ProSci Inc.
91-930
PRSI91-930EA 365 EUR InStock
PRSI91-930
Human recombinant BMP Receptor IA (from Cells)
Protéines et Peptides
Bone Morphogenetic Protein Receptor Type-1A (BMPR1A) belongs to the TKL Ser/Thr protein kinase family and TGFB receptor subfamily, including the type I receptors BMPR1A and BMPR1B and the type II receptor BMPR2. BMPR1A is a single-pass type I membrane protein and highly expressed in skeletal muscle. BMPR1A contains one GS domain and one protein protein kinase domain. BMPR1A is necessary for the extracellular matrix depostition by osteoblasts. BMPR1A can activate SMAD transcriptional regulators, binding with ligands. Defects in BMPR1A are a cause of juvenile polyposis syndrome, Cowden disease and hereditary mixed polyposis syndrome 2 (HMPS2).
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité