Human Recombinant CD27 (from Cells)
Fournisseur: ProSci Inc.
91-931
PRSI91-931EA 377 EUR InStock
PRSI91-931
Human Recombinant CD27 (from Cells)
Protéines et Peptides
CD27 antigen is also known as CD27L receptor, T-cell activation antigen CD27, Tumor necrosis factor receptor superfamily member 7, T14 and TNFRSF7. In humans, it is encoded by the CD27 gene. CD27 is a single-pass type I membrane protein with 3 TNFR-Cys repeats. It is a member of the TNF-receptor superfamily and is required for generation and long-term maintenance of T cell immunity. It binds to ligand CD70, and plays a key role in regulating B-cell activation and immunoglobulin synthesis. It plays a role in survival of activated T-cells and apoptosis through association with SIVA1.
  • This recombinant protein can be used for biological assays. For research use only.
Fusion-Tag: C-Fc-6 His tag
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité