Human recombinant ASM-like phosphodiesterase 3a (from Cells)
Fournisseur: ProSci Inc.
91-932
PRSI91-932EA 761 EUR InStock
PRSI91-932
Human recombinant ASM-like phosphodiesterase 3a (from Cells)
Enzymes
Acid sphingomyelinase-like phosphodiesterase 3a (SMPDL3A ) is a novel liver X receptor (LXR) -regulated gene, with an LXR response element within its promoter. The induction of SMPDL3A is LXR-dependent and is restricted to human blood cells with no induction observed in mouse cellular systems. LXR function as physiological sensors of cholesterol metabolites (oxysterols), regulating key genes involved in cholesterol and lipid metabolism. LXRs have been extensively studied in both human and rodent cell systems, revealing their potential therapeutic value in the contexts of atherosclerosis and inflammatory diseases.
Fusion-Tag: C-6 His tag
This recombinant protein can be used for biological assays. For research use only.
Saisissez le numéro de lot :
Choisissez parmi les numéros de lot/lot récents :
Ou
Produit original :
Description Numéro de catalogue Disponibilité Unité Votre prix Prix par Quantité